4,4-Difluorcyclohexanon, Sigma-Aldrich®

Lieferant: Merck

711403-250MG
SIAL711403-250MGEA 185 EUR
SIAL711403-250MG
4,4-Difluorcyclohexanon, Sigma-Aldrich®
4,4-Difluorocyclohexanone
Formel: C₆H₈F₂O
Molecular Weight: 134,12 g/mol
MDL: MFCD08146635
CAS-Nummer: 22515-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR