6-Chlorchinoxalin, Sigma-Aldrich®

Lieferant: Merck

711411-5G
SIAL711411-5GEA 50.2 EUR
SIAL711411-5G
6-Chlorchinoxalin, Sigma-Aldrich®
6-Chloroquinoxaline
Formel: C₈H₅ClN₂
MDL: MFCD00047594
CAS-Nummer: 5448-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR