5-Iod-1,2,3-trimethoxybenzol, Sigma-Aldrich®

Lieferant: Merck

711535-5G
SIAL711535-5GEA 275 EUR
SIAL711535-5G
5-Iod-1,2,3-trimethoxybenzol, Sigma-Aldrich®
5-Iod-1,2,3-trimethoxybenzol
Formel: C₉H₁₁IO₃
Molecular Weight: 294.09 g/mol
MDL: MFCD01318153
CAS-Nummer: 25245-29-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR