1-Butyl-3-methylimidazoliumtrifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Butyl-3-methylimidazolium Trifluoromethanesulphonate

711772-100G
SIAL711772-100GEA 312 EUR
SIAL711772-100G
1-Butyl-3-methylimidazoliumtrifluormethansulfonat, Sigma-Aldrich®
1-Butyl-3-methylimidazoliumtrifluoromethanesulphonate
Formel: C₉H₁₅F₃N₂O₃S
MDL: MFCD03427620
CAS-Nummer: 174899-66-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR