1-Ethyl-3-methylimidazoliumiodid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-ethyl-3-methylimidazolium iodide

713031-50G 713031-5G
SIAL713031-50GEA 219 EUR
SIAL713031-50G SIAL713031-5G
1-Ethyl-3-methylimidazoliumiodid, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium iodide
Formel: C₆H₁₁IN₂
MDL: MFCD03701101
CAS-Nummer: 35935-34-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR