(Iodmethyl)cyclopropan, Sigma-Aldrich®

Lieferant: Merck

713457-1G
SIAL713457-1GEA 151 EUR
SIAL713457-1G
(Iodmethyl)cyclopropan, Sigma-Aldrich®
(Iodomethyl)cyclopropane
Formel: C₄H₇I
Molecular Weight: 182,00 g/mol
MDL: MFCD06410734
CAS-Nummer: 33574-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR