1-Boc-DL-nipecotinsäureethylester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate, Ethyl 1-Boc-DL-nipecotate, Ethyl 1-Boc-3-piperidinecarboxylate

713724-5G
SIAL713724-5GEA 0 EUR
SIAL713724-5G
1-Boc-DL-nipecotinsäureethylester, Sigma-Aldrich®
1-Boc-DL-nipecotinsäureethylester
Formel: C₁₃H₂₃NO₄
Molecular Weight: 257,33 g/mol
MDL: MFCD04116274
CAS-Nummer: 130250-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR