5-Bromchinolin, Sigma-Aldrich®

Lieferant: Merck
Achtung

714429-5G
SIAL714429-5GEA 163 EUR
SIAL714429-5G
5-Bromchinolin, Sigma-Aldrich®
5-Bromoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,06 g/mol
MDL: MFCD00234481
CAS-Nummer: 4964-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR