3'-Amino-4'-bromacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(3-Amino-4-bromophenyl)ethanone

714437-5G
SIAL714437-5GEA 111 EUR
SIAL714437-5G
3'-Amino-4'-bromacetophenon, Sigma-Aldrich®
3'-Amino-4'-bromacetophenon
Formel: C₈H₈BrNO
Molecular Weight: 214,592 g/mol
MDL: MFCD00466451
CAS-Nummer: 37148-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR