(2-Chlor-4-pyridinyl)methanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2-Chloropyridin-4-yl)methanol, 2-Chloro-4-(hydroxymethyl)pyridine, 2-Chloropyridine-4-methanol

714445-250MG
SIAL714445-250MGEA 35.5 EUR
SIAL714445-250MG
(2-Chlor-4-pyridinyl)methanol, Sigma-Aldrich®
(2-Chlor-4-pyridinyl)methanol
Formel: C₆H₆ClNO
Molecular Weight: 143,57 g/mol
MDL: MFCD06858767
CAS-Nummer: 100704-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR