4-Fluor-2-(trifluormethyl)benzolsulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Fluor-2-(trifluormethyl)benzol-1-sulfonylchlorid, 4-Fluor-2-(trifluormethyl)benzolsulfochlorid, 4-Fluor-2-(trifluormethyl)benzolsulfonsäurechlorid

714496-1G
SIAL714496-1GEA 48.5 EUR
SIAL714496-1G
4-Fluor-2-(trifluormethyl)benzolsulfonylchlorid, Sigma-Aldrich®
4-Fluor-2-(trifluormethyl)benzolsulfonylchlorid
Formel: C₇H₃ClF₄O₂S
Molecular Weight: 262,61 g/mol
MDL: MFCD01091000
CAS-Nummer: 176225-09-5
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR