4-Brom-2,2-difluor-1,3-benzodioxol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 4-Bromo-2,2-difluorobenzo[d][1,3]dioxole

714542-1G
SIAL714542-1GEA 58.5 EUR
SIAL714542-1G
4-Brom-2,2-difluor-1,3-benzodioxol, Sigma-Aldrich®
4-Brom-2,2-difluor-1,3-benzodioxol
Formel: C₇H₃BrF₂O₂
Molecular Weight: 237 g/mol
MDL: MFCD01631385
CAS-Nummer: 144584-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR