2-Chlor-5-(trifluormethyl)benzylalkohol, Sigma-Aldrich®

Lieferant: Merck

714607-5G
SIAL714607-5GEA 31.2 EUR
SIAL714607-5G
2-Chlor-5-(trifluormethyl)benzylalkohol, Sigma-Aldrich®
2-Chlor-5-(trifluormethyl)benzylalkohol
Formel: C₈H₆ClF₃O
Molecular Weight: 210,58 g/mol
MDL: MFCD01631469
CAS-Nummer: 64372-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR