4-Brom-2-fluorbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Brom-2-fluorphenyl)methanol

714615-5G
SIAL714615-5GEA 80.4 EUR
SIAL714615-5G
4-Brom-2-fluorbenzylalkohol, Sigma-Aldrich®
4-Brom-2-fluorbenzylalkohol
Formel: C₇H₆BrFO
Molecular Weight: 205,03 g/mol
MDL: MFCD00143265
CAS-Nummer: 188582-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR