5-Chlor-2,3-diaminopyridin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,3-Diamino-5-chlorpyridin, 5-Chlorpyridin-2,3-diyldiamin
714623-5G
SIAL714623-5GEA
56.5
EUR
SIAL714623-5G
5-Chlor-2,3-diaminopyridin, Sigma-Aldrich®
5-Chlor-2,3-diaminopyridin
Formel:
C₅H₆ClN₃ Molecular Weight: 143.58 g/mol Siedepunkt: 288 Schmelzpunkt: 174…175 °C |
MDL:
MFCD01230986 CAS-Nummer: 25710-20-7 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...