2-Amino-5-chlor-3-fluorpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Chlor-3-fluor-2-pyridylamin, 5-Chlor-3-fluorpyridin-2-amin

714631-1G
SIAL714631-1GEA 76.6 EUR
SIAL714631-1G
2-Amino-5-chlor-3-fluorpyridin, Sigma-Aldrich®
2-Amino-5-chloro-3-fluoropyridine
Formel: C₅H₄ClFN₂
Molecular Weight: 146,55 g/mol
MDL: MFCD09878434
CAS-Nummer: 246847-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR