3-Bromisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Bromopyridine-4-carboxylic acid, 3-Bromo-4-pyridinecarboxylic acid

714658-1G
SIAL714658-1GEA 52.4 EUR
SIAL714658-1G
3-Bromisonicotinsäure, Sigma-Aldrich®
3-Bromisonicotinsäure
Formel: C₆H₄BrNO₂
Molecular Weight: 202.01 g/mol
MDL: MFCD00040944
CAS-Nummer: 13959-02-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR