2-Bromchinolin, Sigma-Aldrich®

Lieferant: Merck
Achtung

714666-1G
SIAL714666-1GEA 33.6 EUR
SIAL714666-1G
2-Bromchinolin, Sigma-Aldrich®
2-Bromoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,06 g/mol
MDL: MFCD00234480
CAS-Nummer: 2005-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR