Isobutyl-4-hydroxybenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Isobutyl 4-hydroxybenzoate, Isobutylparaben

715077-25G
SIAL715077-25GEA 66.7 EUR
SIAL715077-25G
Isobutyl-4-hydroxybenzoat, Sigma-Aldrich®
Isobutyl-4-hydroxybenzoat
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,23 g/mol
MDL: MFCD00020167
CAS-Nummer: 4247-02-3
EINECS: 224-208-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR