2-Fluor-5-formylphenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluoro-5-formylbenzeneboronic acid pinacol ester, 4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 2-Fluoro-5-formylphenylboronic acid, pinacol ester

715328-500MG
SIAL715328-500MGEA 98.7 EUR
SIAL715328-500MG
2-Fluor-5-formylphenylboronsäurepinakolester, Sigma-Aldrich®
2-Fluor-5-formylphenylboronsäurepinakolester
Formel: C₁₃H₁₆BFO₃
Molecular Weight: 250,07 g/mol
MDL: MFCD11867877
CAS-Nummer: 1112208-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR