1H-Indazol-5-boronsäure Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

715409-250MG
SIAL715409-250MGEA 120 EUR
SIAL715409-250MG
1H-Indazol-5-boronsäure Hydrochlorid, Sigma-Aldrich®
1H-Indazol-5-boronsäure Hydrochlorid
Formel: C₇H₈BClN₂O₂
MDL: MFCD11867883
CAS-Nummer: 1257738-46-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR