2-Benzyloxy-5-pyridinboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Benzyloxypyridine-3-boronic acid pinacol ester, 6-Benzyloxypyridine-3-boronic acid, pinacol ester, 2-(Benzyloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

715484-1G
SIAL715484-1GEA 61.4 EUR
SIAL715484-1G
2-Benzyloxy-5-pyridinboronsäurepinakolester, Sigma-Aldrich®
2-Benzyloxy-5-pyridinboronsäurepinakolester
Formel: C₁₈H₂₂BNO₃
Molecular Weight: 311,19 g/mol
MDL: MFCD06798263
CAS-Nummer: 832735-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR