4-(Dimethylcarbamoyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-(Dimethylcarbamoyl)phenyl)boronic acid, 4-(Dimethylcarbamoyl)benzeneboronic acid, 4-(N,N-Dimethylaminocarbonyl)phenylboronic acid

715492-1G
SIAL715492-1GEA 37.6 EUR
SIAL715492-1G
4-(Dimethylcarbamoyl)phenylboronsäure, Sigma-Aldrich®
4-(Dimethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₂BNO₃
Molecular Weight: 193,01 g/mol
MDL: MFCD02258943
CAS-Nummer: 405520-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR