2,3-Dibrom-5-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3-Dibrom-5-methylpyridin

715727-1G
SIAL715727-1GEA 46.4 EUR
SIAL715727-1G
2,3-Dibrom-5-picolin, Sigma-Aldrich®
2,3-Dibrom-5-picolin
Formel: C₆H₅Br₂N
Molecular Weight: 250,92 g/mol
MDL: MFCD04112574
CAS-Nummer: 29232-39-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR