2,2-Difluor-1,3-benzodioxol-4-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2-Difluorobenzo[d][1,3]dioxole-4-carboxylic acid

716359-1G
SIAL716359-1GEA 43.5 EUR
SIAL716359-1G
2,2-Difluor-1,3-benzodioxol-4-carbonsäure, Sigma-Aldrich®
2,2-Difluor-1,3-benzodioxol-4-carbonsäure
Formel: C₈H₄F₂O₄
Molecular Weight: 202,12 g/mol
MDL: MFCD01631473
CAS-Nummer: 126120-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR