2,5-Dibrom-3-methylthiophen, Sigma-Aldrich®

Lieferant: Merck

716375-5G
SIAL716375-5GEA 63.6 EUR
SIAL716375-5G
2,5-Dibrom-3-methylthiophen, Sigma-Aldrich®
2,5-Dibromo-3-methylthiophene
Formel: C₅H₄Br₂S
Molecular Weight: 255,97 g/mol
MDL: MFCD00015470
CAS-Nummer: 13191-36-1
EINECS: 236-147-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR