5-Nitro-2-benzofurancarbonsäure, Sigma-Aldrich®

Lieferant: Merck

716383-1G
SIAL716383-1GEA 41.6 EUR
SIAL716383-1G
5-Nitro-2-benzofurancarbonsäure, Sigma-Aldrich®
5-Nitrobenzofuran-2-carboxylic acid
Formel: C₉H₅NO₅
Molecular Weight: 207,1397 g/mol
MDL: MFCD00060513
CAS-Nummer: 10242-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR