(3-Hydroxy-4-methoxyphenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(3-Hydroxy-4-methoxyphenyl)acetic acid

716391-1G
SIAL716391-1GEA 56.1 EUR
SIAL716391-1G
(3-Hydroxy-4-methoxyphenyl)essigsäure, Sigma-Aldrich®
(3-Hydroxy-4-methoxyphenyl)essigsäure
Formel: (C₆H₃(OH)(OCH₃))CH₂COOH
Molecular Weight: 182,18 g/mol
MDL: MFCD00016829
CAS-Nummer: 1131-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR