4-Iodsalicylsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-4-iodobenzoic acid

716405-5G
SIAL716405-5GEA 122 EUR
SIAL716405-5G
4-Iodsalicylsäure, Sigma-Aldrich®
4-Iodsalicylsäure
Formel: C₇H₅IO₃
Molecular Weight: 264,02 g/mol
MDL: MFCD00061127
CAS-Nummer: 16870-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR