6-Brom-m-toluylsäure, Sigma-Aldrich®

Lieferant: Merck

716413-1G
SIAL716413-1GEA 31.9 EUR
SIAL716413-1G
6-Brom-m-toluylsäure, Sigma-Aldrich®
6-Brom-m-toluylsäure
Formel: C₈H₇BrO₂
Molecular Weight: 215.05 g/mol
MDL: MFCD00079722
CAS-Nummer: 6967-82-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR