4-Bromsalicylsäure, Sigma-Aldrich®

Lieferant: Merck

716421-5G
SIAL716421-5GEA 106 EUR
SIAL716421-5G
4-Bromsalicylsäure, Sigma-Aldrich®
4-Bromsalicylsäure
Formel: C₇H₅BrO₃
MDL: MFCD06797970
CAS-Nummer: 1666-28-0
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR