3-Brom-2-thiophencarbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Bromothiophene-2-carboxaldehyde, 3-Bromo-2-formylthiophene

716553-1G
SIAL716553-1GEA 34.3 EUR
SIAL716553-1G
3-Brom-2-thiophencarbaldehyd, Sigma-Aldrich®
3-Brom-2-thiophencarbaldehyd
Formel: C₅H₃BrOS
Molecular Weight: 191,05 g/mol
MDL: MFCD00126680
CAS-Nummer: 930-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR