4'-Methoxy-2-chloracetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chloro-1-(4-methoxyphenyl)ethanone, 4-Methoxyphenacyl chloride

716618-1G
SIAL716618-1GEA 69.7 EUR
SIAL716618-1G
4'-Methoxy-2-chloracetophenon, Sigma-Aldrich®
4'-Methoxy-2-chloracetophenon
Formel: C₉H₉ClO₂
Molecular Weight: 184,6196 g/mol
MDL: MFCD00216508
CAS-Nummer: 2196-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR