Sodium bitartrate monohydrate ≥99,0% (durch titrimetrische Analyse), BioUltra, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(2R,3R)-2,3-Dihydroxysuccinic acid, sodium salt hydrate
71679-250G
SIAL71679-250GEA
104
EUR
SIAL71679-250G
Sodium bitartrate monohydrate ≥99,0% (durch titrimetrische Analyse), BioUltra, Sigma-Aldrich®
Sodium bitartrate monohydrate
Formel:
C₄H₅NaO₆·H₂O Molecular Weight: 190,083 g/mol |
MDL:
MFCD00150038 CAS-Nummer: 6131-98-2 EINECS: 208-400-9 |
Spezifikation Testergebnisse
Assay | ≥99.0% (T) |
chloride (Cl⁻) | ≤50 mg/kg |
sulphate (SO₄)²⁻ | ≤50 mg/kg |
Al | ≤5 mg/kg |
As | ≤0.1 mg/kg |
Ba | ≤5 mg/kg |
Bi | ≤5 mg/kg |
Ca | ≤50 mg/kg |
Cd | ≤5 mg/kg |
Co | ≤5 mg/kg |
Cr | ≤5 mg/kg |
Cu | ≤5 mg/kg |
Fe | ≤5 mg/kg |
K | ≤50 mg/kg |
Li | ≤5 mg/kg |
MG | ≤5 mg/kg |
Mn | ≤5 mg/kg |
Mo | ≤5 mg/kg |
Ni | ≤5 mg/kg |
PB | ≤5 mg/kg |
Sr | ≤5 mg/kg |
Zn | ≤5 mg/kg |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...