Benzamidoxim, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N'-Hydroxybenzimidamide, Benzamide oxime

717436-1G
SIAL717436-1GEA 31.5 EUR
SIAL717436-1G
Benzamidoxim, Sigma-Aldrich®
Benzamidoxim
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
MDL: MFCD00031485
CAS-Nummer: 613-92-3
EINECS: 210-361-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR