3-Brom-1H-indazol, Sigma-Aldrich®

Lieferant: Merck

717525-1G
SIAL717525-1GEA 35 EUR
SIAL717525-1G
3-Brom-1H-indazol, Sigma-Aldrich®
3-Bromo-1H-indazole
Formel: C₇H₅BrN₂
Molecular Weight: 197,04 g/mol
MDL: MFCD00159926
CAS-Nummer: 40598-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR