(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol, Sigma-Aldrich®
Lieferant: Merck
717746-500MG
SIAL717746-500MGEA
1000
EUR
SIAL717746-500MG
(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol, Sigma-Aldrich®
(S)-3,3'-Di(phenanthren-9-yl)-1,1'-binaphthyl-2,2'-diol
Formel:
C₄₈H₃₀O₂ |
MDL:
MFCD09027270 CAS-Nummer: 361342-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...