2-Amino-4-(4-bromphenyl)-5-methylthiazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(4-Bromophenyl)-5-methylthiazol-2-amine

717851-1G
SIAL717851-1GEA 43.7 EUR
SIAL717851-1G
2-Amino-4-(4-bromphenyl)-5-methylthiazol, Sigma-Aldrich®
2-Amino-4-(4-bromophenyl)-5-methylthiazole
Formel: C₁₀H₉BrN₂S
Molecular Weight: 269,16 g/mol
MDL: MFCD00170268
CAS-Nummer: 65705-44-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR