3-(3-Bromophenyl)-1H-pyrazole, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(3-Bromophenyl)-1H-pyrazole

717878-1G
SIAL717878-1GEA 65 EUR
SIAL717878-1G
3-(3-Bromophenyl)-1H-pyrazole, Sigma-Aldrich®
3-(3-Bromophenyl)-1H-pyrazole
Formel: C₉H₇BrN₂
Molecular Weight: 223,07 g/mol
MDL: MFCD01940433
CAS-Nummer: 149739-65-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR