3-Phenyl-5-isoxazolcarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Phenylisoxazole-5-carboxylic acid

718092-1G
SIAL718092-1GEA 55.8 EUR
SIAL718092-1G
3-Phenyl-5-isoxazolcarbonsäure, Sigma-Aldrich®
3-Phenyl-5-isoxazolecarboxylic acid
Formel: C₁₀H₇NO₃
Molecular Weight: 189,17 g/mol
MDL: MFCD01936010
CAS-Nummer: 14442-12-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR