2,3-Difluorpyridin, Sigma-Aldrich®

Lieferant: Merck
Gefahr

718173-5G
SIAL718173-5GEA 82.9 EUR
SIAL718173-5G
2,3-Difluorpyridin, Sigma-Aldrich®
2,3-Difluoropyridine
Formel: C₅H₃F₂N
Molecular Weight: 115.08 g/mol
MDL: MFCD04114143
CAS-Nummer: 1513-66-2
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR