1-Methyl-2-imidazolsulfonylchlorid, Sigma-Aldrich®

Lieferant: Merck

718211-1G
SIAL718211-1GEA 182 EUR
SIAL718211-1G
1-Methyl-2-imidazolsulfonylchlorid, Sigma-Aldrich®
1-Methyl-2-imidazolsulfonylchlorid
Formel: C₄H₅ClN₂O₂S
MDL: MFCD16621450
CAS-Nummer: 55694-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR