4-Oxazolcarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Oxazole-4-carboxylic acid

718343-1G
SIAL718343-1GEA 124 EUR
SIAL718343-1G
4-Oxazolcarbonsäure, Sigma-Aldrich®
4-Oxazolecarboxylic acid
Formel: C₄H₃NO₃
Molecular Weight: 113,0715 g/mol
MDL: MFCD06797151
CAS-Nummer: 23012-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR