2-Methyl-4-thiazolcarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methylthiazole-4-carboxylic acid

718440-1G
SIAL718440-1GEA 58.6 EUR
SIAL718440-1G
2-Methyl-4-thiazolcarbonsäure, Sigma-Aldrich®
2-Methyl-4-thiazolecarboxylic acid
Formel: C₅H₅NO₂S
Molecular Weight: 143,1637 g/mol
MDL: MFCD03407332
CAS-Nummer: 35272-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR