2-Chlor-3-cyan-4-pyridinboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chloro-3-cyanopyridine-4-boronic acid pinacol ester

718920-1G
SIAL718920-1GEA 540 EUR
SIAL718920-1G
2-Chlor-3-cyan-4-pyridinboronsäurepinakolester, Sigma-Aldrich®
2-Chlor-3-cyan-4-pyridinboronsäurepinakolester
Formel: C₁₂H₁₄BClN₂O₂
Molecular Weight: 264.52 g/mol
MDL: MFCD09037479
CAS-Nummer: 878194-94-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR