1-Methyl-2-(tributylstannyl)indol, Sigma-Aldrich®

Lieferant: Merck
Gefahr

719366-1G
SIAL719366-1GEA 247 EUR
SIAL719366-1G
1-Methyl-2-(tributylstannyl)indol, Sigma-Aldrich®
1-Methyl-2-(tributylstannyl)indol
Formel: C₂₁H₃₅NSn
Molecular Weight: 420,21 g/mol
MDL: MFCD09265483
CAS-Nummer: 157427-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR