4'-Brombiphenyl-3-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4'-Bromo-[1,1'-biphenyl]-3-carboxylic acid

720097-1G
SIAL720097-1GEA 80.5 EUR
SIAL720097-1G
4'-Brombiphenyl-3-carbonsäure, Sigma-Aldrich®
4'-Brombiphenyl-3-carbonsäure
Formel: C₁₃H₉BrO₂
Molecular Weight: 277,12 g/mol
MDL: MFCD01318398
CAS-Nummer: 885951-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR