(2-Methyl-3-nitrophenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methyl-3-nitrophenylacetic Acid

720216-5G
SIAL720216-5GEA 28.6 EUR
SIAL720216-5G
(2-Methyl-3-nitrophenyl)essigsäure, Sigma-Aldrich®
(2-Methyl-3-nitrophenyl)essigsäure
Formel: C₉H₉NO₄
Molecular Weight: 195,17 g/mol
MDL: MFCD07782132
CAS-Nummer: 23876-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR