2-Amino-4-brompyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Brompyridin-2-amin, 4-Brom-2-pyridylamin

720224-1G
SIAL720224-1GEA 33.3 EUR
SIAL720224-1G
2-Amino-4-brompyridin, Sigma-Aldrich®
2-Amino-4-bromopyridine
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
MDL: MFCD01646115
CAS-Nummer: 84249-14-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR