2-Amino-3-chlorpyrazin ≥97%, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: 3-Chloropyrazin-2-amine, 3-Chloro-2-pyrazinamine

720364-1G
SIAL720364-1GEA 26 EUR
SIAL720364-1G
2-Amino-3-chlorpyrazin ≥97%, Sigma-Aldrich®
2-Amino-3-chloropyrazine
Formel: C₄H₄ClN₃
Molecular Weight: 129,55 g/mol
Schmelzpunkt: 164-169 °C
MDL: MFCD04114305
CAS-Nummer: 6663-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR